1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,6-Dichloro-1-fluoropyridin-1-ium tetrafluoroborate, 2,6-Dichloro-N-fluoropyridinium tetrafluoroborate
M-Clarity™ Qualitätsstufe = MQ100
439290-1G
439290-5G
SIAL439290-1GEA
129
EUR
SIAL439290-1G
SIAL439290-5G
1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat, Sigma-Aldrich®
1-Fluor-2,6-dichlorpyridiniumtetrafluoroborat
Formel:
C₅H₃BCl₂F₅N Molecular Weight: 253,793 g/mol |
MDL:
MFCD00153157 CAS-Nummer: 140623-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...