N-Boc-3-brom-5-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

AN-1375-5G AN-1375-25G AN-1375-1G
COBBAN-1375-5GEA 0 EUR
COBBAN-1375-5G COBBAN-1375-25G COBBAN-1375-1G
N-Boc-3-brom-5-(trifluormethyl)anilin
N-Boc-3-brom-5-(trifluormethyl)anilin
Formel: C₁₂H₁₃BrF₃NO₂
MDL: MFCD11504832
CAS-Nummer: 641571-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR