N-Boc-2-fluor-4-iodanilin

Lieferant: COMBI-BLOCKS

AN-1137-5G AN-1137-1G
COBBAN-1137-5GEA 188 EUR
COBBAN-1137-5G COBBAN-1137-1G
N-Boc-2-fluor-4-iodanilin
N-Boc-2-fluor-4-iodanilin
Formel: C₁₁H₁₃FINO₂
MDL: MFCD06797742
CAS-Nummer: 886497-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR