(2,4-Dichlorphenyl)methansulfonsäurechlorid
Lieferant: CHEMPUR
Synonyme:
2,4-Dichlorobenzylsulphonyl chloride
FL13514.POR
013514-5G
013514-1G
CHMPFL13514.POREA
0
EUR
CHMPFL13514.POR
FLUO013514-5G
FLUO013514-1G
(2,4-Dichlorphenyl)methansulfonsäurechlorid
(2,4-Dichlorophenyl)methanesulphonyl chloride
Formel:
C₇H₅CI₃O₂S |
MDL:
MFCD04116370 CAS-Nummer: 88691-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...