(2-Chlorphenyl)methansulfonsäurechlorid

Lieferant: CHEMPUR

FL13481.POR 020876-1G
CHMPFL13481.POREA 0 EUR
CHMPFL13481.POR FLUO020876-1G
(2-Chlorphenyl)methansulfonsäurechlorid
(2-Chlorophenyl)methanesulphonyl chloride
Formel: C₇H₆Cl₂O₂S
Molecular Weight: 225.923 g/mol
MDL: MFCD04116369
CAS-Nummer: 77421-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR