(3,5-Dichlorphenyl)methansulfonsäurechlorid 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3,5-Dichloro-α-toluenesulphonyl chloride

H50707.03 H50707.MD
H50707.03EA 134 EUR
H50707.03 H50707.MD
(3,5-Dichlorphenyl)methansulfonsäurechlorid 96%
(3,5-Dichlorophenyl)methanesulphonyl chloride

Purity: 96% CAS No.: 163295-70-3 Molecular Formula: C7H5Cl3O2S Formula Weight: 259.54 UN No.: 3096 MDL No.: MFCD01631928

Formel: C₇H₅CI₃O₂S
Molecular Weight: 259.54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631928
CAS-Nummer: 163295-70-3
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR