(4-Chlorphenyl)methansulfonsäurechlorid

Lieferant: CHEMPUR

Synonyme: 4-Chloro-α-toluenesulphonyl chloride

FL13482.POR 020874-1G 020874-5G
CHMPFL13482.POREA 0 EUR
CHMPFL13482.POR FLUO020874-1G FLUO020874-5G
(4-Chlorphenyl)methansulfonsäurechlorid
(4-Chlorophenyl)methanesulphonyl chloride
Formel: C₇H₆Cl₂O₂S
Molecular Weight: 225,10 g/mol
MDL: MFCD01631927
CAS-Nummer: 6966-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR