(3-Nitrophenyl)methansulfonsäurechlorid 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Nitro-α-toluenesulphonyl chloride

B23333.06 B23333.03 B23333.14
B23333.06EA 134 EUR
B23333.06 B23333.03 B23333.14
(3-Nitrophenyl)methansulfonsäurechlorid 95%
(3-Nitrophenyl)methanesulphonyl chloride

Purity: 95% CAS No.: 58032-84-1 Molecular Formula: C7H6ClNO4S Formula Weight: 235.64 Melting Point: 95-102° UN No.: 3261 MDL No.: MFCD01859935

Formel: C₇H₆ClNO₄S
Molecular Weight: 235.64 g/mol
Schmelzpunkt: 95…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01859935
CAS-Nummer: 58032-84-1
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR