4-(Methylsulfonyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid

M-Clarity™ Qualitätsstufe = MQ100

675903-1G 675903-5G
SIAL675903-1GEA 37.4 EUR
SIAL675903-1G SIAL675903-5G
4-(Methylsulfonyl)phenylboronsäure, Sigma-Aldrich®
4-(Methylsulfonyl)phenylboronsäure
Formel: C₇H₉BO₄S
Molecular Weight: 200,02 g/mol
MDL: MFCD01630820
CAS-Nummer: 149104-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR