2-Brom-N-methyl-5-(trifluormethoxy)anilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2926-1G HC-2926-5G
COBBHC-2926-1GEA 204 EUR
COBBHC-2926-1G COBBHC-2926-5G
2-Brom-N-methyl-5-(trifluormethoxy)anilin Hydrochlorid
2-Brom-N-methyl-5-(trifluormethoxy)anilin Hydrochlorid
Formel: C₈H₈BrClF₃NO
MDL: MFCD15143545
CAS-Nummer: 1215206-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR