2-Brom-4-thiocyanatoanilin

Lieferant: COMBI-BLOCKS

AN-1397-5G AN-1397-1G
COBBAN-1397-5GEA 303 EUR
COBBAN-1397-5G COBBAN-1397-1G
2-Brom-4-thiocyanatoanilin
2-Brom-4-thiocyanatoanilin
Formel: C₇H₅BrN₂S
MDL: MFCD04618254
CAS-Nummer: 7493-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR