2-(2,4-Dichlorphenoxy)anilin Hydrochlorid
Lieferant: Thermo Scientific
SB00593.1
SB00593.10
SB00593.50
MAYBSB00593.1EA
0
EUR
MAYBSB00593.1
MAYBSB00593.10
MAYBSB00593.50
2-(2,4-Dichlorphenoxy)anilin Hydrochlorid
2-(2,4-Dichlorphenoxy)anilin Hydrochlorid
Formel:
C₁₂H₁₀Cl₃NO Molecular Weight: 290,58 g/mol |
MDL:
MFCD00052001 CAS-Nummer: 89279-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...