2,3-Dichlor-4-thiocyanatoanilin

Lieferant: COMBI-BLOCKS

HC-2729-5G HC-2729-1G
COBBHC-2729-5GEA 303 EUR
COBBHC-2729-5G COBBHC-2729-1G
2,3-Dichlor-4-thiocyanatoanilin
2,3-Dichlor-4-thiocyanatoanilin
Formel: C₇H₄Cl₂N₂S
MDL: MFCD11556361
CAS-Nummer: 7494-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR