3-Chlor-4-thiocyanatoanilin

Lieferant: COMBI-BLOCKS

AN-1406-5G AN-1406-1G
COBBAN-1406-5GEA 303 EUR
COBBAN-1406-5G COBBAN-1406-1G
3-Chlor-4-thiocyanatoanilin
3-Chlor-4-thiocyanatoanilin
Formel: C₇H₅ClN₂S
MDL: MFCD07613584
CAS-Nummer: 3226-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR