3-Fluor-4-thiocyanatoanilin

Lieferant: COMBI-BLOCKS

AN-1416-5G AN-1416-1G
COBBAN-1416-5GEA 451 EUR
COBBAN-1416-5G COBBAN-1416-1G
3-Fluor-4-thiocyanatoanilin
3-Fluor-4-thiocyanatoanilin
Formel: C₇H₅FN₂S
MDL: MFCD11855824
CAS-Nummer: 14512-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR