4-Brom-N-methyl-2-(trifluormethyl)anilin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-2878-5G
HC-2878-1G
COBBHC-2878-5GEA
451
EUR
COBBHC-2878-5G
COBBHC-2878-1G
4-Brom-N-methyl-2-(trifluormethyl)anilin Hydrochlorid
4-Brom-N-methyl-2-(trifluormethyl)anilin Hydrochlorid
Formel:
C₈H₈BrClF₃N |
MDL:
MFCD13195689 CAS-Nummer: 1215206-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...