Tetrabutylammoniumtoluol-4-sulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tetrabutylammonium p-Toluenesulphonate, Tetrabutylammonium 4-methylbenzenesulphonate
M-Clarity™ Qualitätsstufe = MQ200
358681-10G
358681-2G
SIAL358681-10GEA
238
EUR
SIAL358681-10G
SIAL358681-2G
Tetrabutylammoniumtoluol-4-sulfonat, Sigma-Aldrich®
Tetrabutylammoniumtoluol-4-sulfonat
Formel:
C₂₃H₄₃NO₃S Molecular Weight: 413.6574 g/mol |
MDL:
MFCD00043227 CAS-Nummer: 7182-86-7 EINECS: 230-548-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...