N-(3,5-Dichlorbenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
Lieferant: Thermo Scientific
RF04579.250
RF04579.1
MAYBRF04579.250EA
0
EUR
MAYBRF04579.250
MAYBRF04579.1
N-(3,5-Dichlorbenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
N-(3,5-Dichlorbenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
Formel:
C₂₀H₁₀Cl₂F₁₃NO |
MDL:
MFCD02180606 CAS-Nummer: 680213-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...