N-(4-Chlorbenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid

Lieferant: Thermo Scientific

RF04577.250 RF04577.1
MAYBRF04577.250EA 0 EUR
MAYBRF04577.250 MAYBRF04577.1
N-(4-Chlorbenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
N-(4-Chlorbenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
Formel: C₂₀H₁₀Cl₂F₁₃NO
MDL: MFCD02180604
CAS-Nummer: 680213-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR