N-(4-Methoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid

Lieferant: Thermo Scientific

RF04573.250 RF04573.1
MAYBRF04573.250EA 0 EUR
MAYBRF04573.250 MAYBRF04573.1
N-(4-Methoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
N-(4-Methoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
Formel: C₂₁H₁₄F₁₃NO₂
MDL: MFCD00179695
CAS-Nummer: 288161-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR