N-(2,6-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid

Lieferant: Thermo Scientific

RF04576.250 RF04576.1
MAYBRF04576.250EA 0 EUR
MAYBRF04576.250 MAYBRF04576.1
N-(2,6-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
N-(2,6-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorhexyl)benzamid
Formel: C₂₂H₁₆F₁₃NO₃
MDL: MFCD02180603
CAS-Nummer: 680213-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR