2,3-Dimethyl-4-thiocyanatoanilin

Lieferant: COMBI-BLOCKS

AN-1405-5G AN-1405-1G
COBBAN-1405-5GEA 303 EUR
COBBAN-1405-5G COBBAN-1405-1G
2,3-Dimethyl-4-thiocyanatoanilin
2,3-Dimethyl-4-thiocyanatoanilin
Formel: C₉H₁₀N₂S
MDL: MFCD11855820
CAS-Nummer: 23530-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR