N-Benzylbenzamid

Lieferant: MENAI

IP9.100 IP9.500
MENAIP9.100EA 0 EUR
MENAIP9.100 MENAIP9.500
N-Benzylbenzamid
N-Benzylbenzamid
Formel: C₁₄H₁₃NO
MDL: MFCD00003070
CAS-Nummer: 1485-70-7
EINECS: 216-063-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR