4-Carboxy-1-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Borono-1-naphthoic acid

BB-3908-1G BB-3908-250M
COBBBB-3908-1GEA 864 EUR
COBBBB-3908-1G COBBBB-3908-250M
4-Carboxy-1-naphthalinboronsäure
4-Carboxy-1-naphthalinboronsäure
Formel: C₁₁H₉BO₄
Molecular Weight: 216.00 g/mol
MDL: MFCD06203538
CAS-Nummer: 332398-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR