2-Ethoxy-1-naphthalinboronsäure
Lieferant: COMBI-BLOCKS
BB-8196-1G
BB-8196-250M
COBBBB-8196-1GEA
38.2
EUR
COBBBB-8196-1G
COBBBB-8196-250M
2-Ethoxy-1-naphthalinboronsäure
2-Ethoxy-1-naphthaleneboronic acid
Formel:
C₁₂H₁₃BO₃ Molecular Weight: 216.0408 g/mol |
MDL:
MFCD05664211 CAS-Nummer: 148345-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...