2-Methoxy-1-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

FA-2314-5G FA-2314-1G
COBBFA-2314-5GEA 548 EUR
COBBFA-2314-5G COBBFA-2314-1G
2-Methoxy-1-naphthalinboronsäure
2-Methoxy-1-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₃
Molecular Weight: 202.0100 g/mol
MDL: MFCD11044173
CAS-Nummer: 104116-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR