3-Methoxy-2-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methoxynaphthalen-2-boronic acid, (3-Methoxynaphthalen-2-yl)boronic acid

BB-5622-5G BB-5622-1G BB-5622-250M
COBBBB-5622-5GEA 933 EUR
COBBBB-5622-5G COBBBB-5622-1G COBBBB-5622-250M
3-Methoxy-2-naphthalinboronsäure
3-Methoxy-2-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₃
Molecular Weight: 202.0100 g/mol
MDL: MFCD12025968
CAS-Nummer: 104115-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR