6-Ethoxy-2-naphthalinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(6-Ethoxynaphthalen-2-yl)boronic acid
BB-3517-1G
BB-3517-25G
BB-3517-100G
BB-3517-5G
COBBBB-3517-1GEA
25.9
EUR
COBBBB-3517-1G
COBBBB-3517-25G
COBBBB-3517-100G
COBBBB-3517-5G
6-Ethoxy-2-naphthalinboronsäure
6-Ethoxy-2-naphthaleneboronic acid
Formel:
C₁₂H₁₃BO₃ Molecular Weight: 216.04 g/mol |
MDL:
MFCD03427009 CAS-Nummer: 352525-98-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...