6-Methoxy-2-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methoxy-2-naphthylboronic acid, 6-Methoxy-2-naphthylboronic Acid

BB-2437-5G BB-2437-25G BB-2437-1G BB-2437-100G
COBBBB-2437-5GEA 71.2 EUR
COBBBB-2437-5G COBBBB-2437-25G COBBBB-2437-1G COBBBB-2437-100G
6-Methoxy-2-naphthalinboronsäure
6-Methoxy-2-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₃
Molecular Weight: 202.02 g/mol
MDL: MFCD03093087
CAS-Nummer: 156641-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR