6-Hydroxy-2-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Hydroxynaphthalen-2-yl)boronic acid, 6-Hydroxynaphthalene-2-boronic acid

BB-2042-1G BB-2042-5G
COBBBB-2042-1GEA 196 EUR
COBBBB-2042-1G COBBBB-2042-5G
6-Hydroxy-2-naphthalinboronsäure
6-Hydroxy-2-naphthaleneboronic acid
Formel: C₁₀H₉BO₃
Molecular Weight: 187.99 g/mol
MDL: MFCD00092986
CAS-Nummer: 173194-95-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR