6-tert-Butyldimethylsilyloxy-2-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-((tert-Butyldimethylsilyl)oxy)naphthalen-2-yl)boronic acid

BB-2041-5G BB-2041-1G
COBBBB-2041-5GEA 451 EUR
COBBBB-2041-5G COBBBB-2041-1G
6-tert-Butyldimethylsilyloxy-2-naphthalinboronsäure
6-tert-Butyldimethylsilyloxy-2-naphthalinboronsäure
Formel: C₁₆H₂₃BO₃Si
Molecular Weight: 302.2485 g/mol
MDL: MFCD04115644
CAS-Nummer: 179942-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR