4-Fluor-1-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoronaphtalene-1-boronic acid, 4-Fluoronaphthalene-1-boronic acid

BB-3556-5G BB-3556-1G
COBBBB-3556-5GEA 658 EUR
COBBBB-3556-5G COBBBB-3556-1G
4-Fluor-1-naphthalinboronsäure
4-Fluoro-1-naphthaleneboronic acid
Formel: C₁₀H₇BBrFO₂
Molecular Weight: 189.98 g/mol
MDL: MFCD04039221
CAS-Nummer: 182344-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR