4-Methyl-1-naphthalinboronsäure 96%
Lieferant: Thermo Scientific
|
Achtung
|
H52998.03
H52998.06
H52998.03EA
41.8
EUR
H52998.03
H52998.06
4-Methyl-1-naphthalinboronsäure 96%
4-Methyl-1-naphthaleneboronic acid
Purity: 96%
CAS No.: 103986-53-4
Molecular Formula: C11H11BO2
Formula Weight: 186.02
Melting Point: 239-240°
MDL No.: MFCD01632204
Formel:
C₁₁H₁₁BO₂ Molecular Weight: 186,02 g/mol Schmelzpunkt: 239…240 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01632204 CAS-Nummer: 103986-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...