4-Methyl-1-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

BB-2463-5G BB-2463-25G BB-2463-1G
COBBBB-2463-5GEA 114 EUR
COBBBB-2463-5G COBBBB-2463-25G COBBBB-2463-1G
4-Methyl-1-naphthalinboronsäure
4-Methyl-1-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₂
Molecular Weight: 186.02 g/mol
MDL: MFCD01632204
CAS-Nummer: 103986-53-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR