1-Phenyl-3-buten-1-ol ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Phenyl-3-buten-1-ol

H53466.06 H53466.14
H53466.06EA 92.4 EUR
H53466.06 H53466.14
1-Phenyl-3-buten-1-ol ≥97%
4-Phenyl-1-buten-4-ol

Purity: 97% CAS No.: 936-58-3 Molecular Formula: C10H12O Formula Weight: 148.20 Flash Point: 103°(217°F) Refractive Index: 1.53 Boiling Point: 234° MDL No.: MFCD00039617

Formel: C₁₀H₁₂O
Molecular Weight: 148,20 g/mol
Siedepunkt: 234 °C
Dichte: 0,992
Flammpunkt: 103 °C (217 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039617
CAS-Nummer: 936-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR