Tetrabutylammoniumfluorid Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrabutylammonium fluoride xhydrate

M-Clarity™ Qualitätsstufe = MQ200

241512-100G 241512-10G
SIAL241512-100GEA 374 EUR
SIAL241512-100G SIAL241512-10G
Tetrabutylammoniumfluorid Hydrat, Sigma-Aldrich®
Tetrabutylammoniumfluorid Hydrat
Formel: C₁₆H₃₆FN·H₂O
Molecular Weight: 279,4774 g/mol
MDL: MFCD00149980
CAS-Nummer: 22206-57-1
EINECS: 207-057-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR