(4-Methylphenyl)methansulfonsäurechlorid
Lieferant: CHEMPUR
Synonyme:
p-Tolylmethanesulphonyl chloride, α-p-Xylenesulphonyl chloride, 4-Methylbenzylsulphonyl chloride
MO00937.10
MO00937.50
CHMPMO00937.10EA
0
EUR
CHMPMO00937.10
CHMPMO00937.50
(4-Methylphenyl)methansulfonsäurechlorid
(4-Methylphenyl)methanesulphonyl chloride
Formel:
C₈H₉ClO₂S Molecular Weight: 204.67 g/mol |
MDL:
MFCD01631931 CAS-Nummer: 51419-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...