3-Fluor-4-iodanilin

Lieferant: COMBI-BLOCKS

HC-3449-5G HC-3449-10G
COBBHC-3449-5GEA 63.9 EUR
COBBHC-3449-5G COBBHC-3449-10G
3-Fluor-4-iodanilin
3-Fluor-4-iodanilin
Formel: C₆H₅FIN
Molecular Weight: 237,01 g/mol
MDL: MFCD07774187
CAS-Nummer: 656-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR