3-Brom-4-fluorbiphenyl

Lieferant: Thermo Scientific

Synonyme: 3-Bromo-4-fluoro-1,1'-biphenyl

SB02017.1 SB02017EA
MAYBSB02017.1EA 0 EUR
MAYBSB02017.1 MAYBSB02017EA
3-Brom-4-fluorbiphenyl
3-Brom-4-fluorbiphenyl
Formel: C₁₂H₈BrF
Molecular Weight: 251,10 g/mol
MDL: MFCD01571091
CAS-Nummer: 306935-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR