1-Ethyl-6-fluorbenzimidazol

Lieferant: COMBI-BLOCKS

IN-0198-5G IN-0198-1G
COBBIN-0198-5GEA 375 EUR
COBBIN-0198-5G COBBIN-0198-1G
1-Ethyl-6-fluorbenzimidazol
1-Ethyl-6-fluorobenzimidazole
Formel: C₉H₉FN₂
MDL: MFCD12546504
CAS-Nummer: 1187385-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR