2-Ethoxy-4-fluoranilin Hydrochlorid

Lieferant: COMBI-BLOCKS

AN-1037-1G AN-1037-5G
COBBAN-1037-1GEA 0 EUR
COBBAN-1037-1G COBBAN-1037-5G
2-Ethoxy-4-fluoranilin Hydrochlorid
2-Ethoxy-4-fluoranilin Hydrochlorid
Formel: C₈H₁₁ClFNO
MDL: MFCD04115636
CAS-Nummer: 850568-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR