4-Fluor-m-anisidin Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-3-methoxyaniline hydrochloride

AN-1071-5G AN-1071-1G
COBBAN-1071-5GEA 595 EUR
COBBAN-1071-5G COBBAN-1071-1G
4-Fluor-m-anisidin Hydrochlorid
4-Fluor-m-anisidin Hydrochlorid
Formel: C₇H₉ClFNO
Molecular Weight: 177,6000 g/mol
MDL: MFCD08689481
CAS-Nummer: 22510-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR