2,5-Dibrom-3,4,6-trifluoranilin

Lieferant: FLUOROCHEM

010290-1G 010290-5G
FLUO010290-1GEA 0 EUR
FLUO010290-1G FLUO010290-5G
2,5-Dibrom-3,4,6-trifluoranilin
2,5-Dibrom-3,4,6-trifluoranilin
Formel: C₆H₂Br₂F₃N
MDL: MFCD01862003
CAS-Nummer: 232267-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR