3-Brom-5-methylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

AN-1065-1G AN-1065-5G
COBBAN-1065-1GEA 30.2 EUR
COBBAN-1065-1G COBBAN-1065-5G
3-Brom-5-methylanilin Hydrochlorid
3-Brom-5-methylanilin Hydrochlorid
Formel: C₇H₉BrClN
MDL: MFCD09475832
CAS-Nummer: 957034-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR