4-Fluor-3-methoxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(4-Fluoro-3-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Fluoro-3-methoxybenzeneboronic acid pinacol ester

PN-2809-5G PN-2809-1G
COBBPN-2809-5GEA 813 EUR
COBBPN-2809-5G COBBPN-2809-1G
4-Fluor-3-methoxyphenylboronsäurepinakolester
4-Fluor-3-methoxyphenylboronsäurepinakolester
Formel: C₁₃H₁₈BFO₃
Molecular Weight: 252,09 g/mol
MDL: MFCD11227055
CAS-Nummer: 425378-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR