1-Isobutyl-1H-pyrazol-4-boronsäurepinakolester
Lieferant: FLUOROCHEM
021615-1G
021615-5G
FLUO021615-1GEA
138
EUR
FLUO021615-1G
FLUO021615-5G
1-Isobutyl-1H-pyrazol-4-boronsäurepinakolester
Formel:
C₁₃H₂₃BN₂O₂ |
MDL:
MFCD05663857 CAS-Nummer: 827614-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...