1-Isopentyl-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS

BB-8531-5G BB-8531-1G BB-8531-250M
COBBBB-8531-5GEA 915 EUR
COBBBB-8531-5G COBBBB-8531-1G COBBBB-8531-250M
1-Isopentyl-1H-pyrazol-4-boronsäure
1-Isopentyl-1H-pyrazol-4-boronsäure
Formel: C₈H₁₅BN₂O₂
MDL: MFCD07781209
CAS-Nummer: 847818-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR