(3-(Trifluormethyl)phenyl)methansulfonsäurechlorid
Lieferant: Thermo Scientific
Synonyme:
3-(Trifluoromethyl)benzylsulphonyl chloride
MO00932CB
MO00932DA
MAYBMO00932CBEA
0
EUR
MAYBMO00932CB
MAYBMO00932DA
(3-(Trifluormethyl)phenyl)methansulfonsäurechlorid
(3-(Trifluoromethyl)phenyl)methanesulphonyl chloride
Formel:
C₈H₆ClF₃O₂S |
MDL:
MFCD01631926 CAS-Nummer: 127162-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...