2-Brom-N-methylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

AN-1181-5G AN-1181-1G
COBBAN-1181-5GEA 257 EUR
COBBAN-1181-5G COBBAN-1181-1G
2-Brom-N-methylanilin Hydrochlorid
2-Brom-N-methylanilin Hydrochlorid
Formel: C₇H₉BrClN
MDL: MFCD09800853
CAS-Nummer: 957120-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR