4-Brom-N-ethylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-3104-5G HC-3104-1G
COBBHC-3104-5GEA 451 EUR
COBBHC-3104-5G COBBHC-3104-1G
4-Brom-N-ethylanilin Hydrochlorid
4-Brom-N-ethylanilin Hydrochlorid
Formel: C₈H₁₁BrClN
MDL: MFCD13196470
CAS-Nummer: 855949-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR